Tisk…

4-(tert-Butoxycarbonyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(tert-Butoxycarbonyl)benzeneboronic acid, (4-(tert-Butoxycarbonyl)phenyl)boronic acid, 4-tert-Butoxycarbonylphenylboronic acid

OR10392-1G OR10392-5G OR10392-25G
APOSOR10392-1GEA 622 CZK
APOSOR10392-1G APOSOR10392-5G APOSOR10392-25G
4-(tert-Butoxycarbonyl)phenylboronic acid 98%
4-(tert-Butoxycarbonyl)phenylboronic acid
Vzorec: C₁₁H₁₅BO₄
MW: 222,05 g/mol
Bod tání: 100…106 °C
Storage Temperature: Ambient
MDL Number: MFCD03411946
CAS číslo: 850568-54-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR