2-(Bromomethyl)phenylboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR10377-1G
OR10377-5G
APOSOR10377-1GEA
622
CZK
APOSOR10377-1G
APOSOR10377-5G
2-(Bromomethyl)phenylboronic acid 95%
2-(Bromomethyl)phenylboronic acid
Vzorec:
C₇H₈BBrO₂ Bod tání: 145…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318941 CAS číslo: 91983-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...