4-Bromoimidazole
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-1H-imidazole
OR10376-1G
OR10376-25G
OR10376-5G
APOSOR10376-1GEA
622
CZK
APOSOR10376-1G
APOSOR10376-25G
APOSOR10376-5G
4-Bromoimidazole
4-Bromoimidazole
Vzorec:
C₃H₃BrN₂ MW: 146,97 g/mol Bod tání: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047021 CAS číslo: 2302-25-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...