2-Bromo-1-methylimidazole 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-1-methyl-1H-imidazole
OR10367-1G
OR10367-5G
APOSOR10367-1GEA
1740
CZK
APOSOR10367-1G
APOSOR10367-5G
2-Bromo-1-methylimidazole 95%
2-Bromo-1-methylimidazole
Vzorec:
C₄H₅BrN₂ MW: 161 g/mol Bod varu: 172 °C Hustota: 1,649 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD02179525 CAS číslo: 16681-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...