Tisk…

2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole

Dodavatel: Apollo Scientific
Varování

OR103663-1G OR103663-5G OR103663-10G
APOSOR103663-1GEA 2980 CZK
APOSOR103663-1G APOSOR103663-5G APOSOR103663-10G
2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole
2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole
Vzorec: C₁₀H₉ClN₂O₂
Storage Temperature: Ambient
CAS číslo: 24023-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR