Tisk…

(4-Boc-aminophenyl)boronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Boc-Aminophenyl)Boronic Acid, 4-Boc-aminophenylboronic acid, (4-Boc-Aminophenyl)boronic acid, 4-(N-BOC-amino)phenylboronic acid, [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic Acid

OR10347-1G OR10347-25G OR10347-5G
APOSOR10347-1GEA 622 CZK
APOSOR10347-1G APOSOR10347-25G APOSOR10347-5G
(4-Boc-aminophenyl)boronic acid 98%
(4-Boc-aminophenyl)boronic acid
Vzorec: C₁₁H₁₆BNO₄
MW: 237,062 g/mol
Bod tání: 199…204 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD02093054
CAS číslo: 380430-49-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR