Tisk…

4-Aminophenylboronic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Aminobenzeneboronic acid hydrochloride, (4-Aminophenyl)boronic acid hydrochloride

OR10338-1G OR10338-25G OR10338-5G
APOSOR10338-1GEA 827 CZK
APOSOR10338-1G APOSOR10338-25G APOSOR10338-5G
4-Aminophenylboronic acid hydrochloride
4-Aminophenylboronic acid hydrochloride
Vzorec: C₆H₉BClNO₂
MW: 173,41 g/mol
Bod tání: 141…143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001333
CAS číslo: 80460-73-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR