Tisk…

3-Aminophenylboronic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Aminophenyl)boronic acid hydrochloride, 3-Aminobenzeneboronic acid hydrochloride

OR10337-1G OR10337-5G OR10337-25G
APOSOR10337-1GEA 622 CZK
APOSOR10337-1G APOSOR10337-5G APOSOR10337-25G
3-Aminophenylboronic acid hydrochloride
3-Aminophenylboronic acid hydrochloride
Vzorec: C₆H₉BClNO₂
Bod tání: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191748
CAS číslo: 85006-23-1
EINECS: 285-052-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR