3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR103361-1G
OR103361-250MG
APOSOR103361-1GEA
953
CZK
APOSOR103361-1G
APOSOR103361-250MG
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Vzorec:
C₁₀H₉ClN₂O₂ MW: 224,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00084966 CAS číslo: 175205-62-6 UN: 1759 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...