Tisk…
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
Dodavatel: Apollo Scientific
Varování
Synonyms: 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine hydrochloride, 4-Aminomethylphenylboronic acid pinacol ester HCl
OR10332-1G OR10332-250MG
APOSOR10332-1GEA 4960 CZK
APOSOR10332-1G APOSOR10332-250MG
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
Vzorec: C₁₃H₂₀BNO₂·HCl
MW: 269.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179455
CAS číslo: 850568-55-7