Tisk…

4-Amino-2-chloro-6,7-dimethoxyquinazoline

Dodavatel: Apollo Scientific
Varování

Synonyms: Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline

OR10316-25G OR10316-5G
APOSOR10316-25GEA 705 CZK
APOSOR10316-25G APOSOR10316-5G
4-Amino-2-chloro-6,7-dimethoxyquinazoline
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Vzorec: C₁₀H₁₀ClN₃O₂
MW: 239,66 g/mol
Bod tání: 262…268 °C
Storage Temperature: Ambient
MDL Number: MFCD00051734
CAS číslo: 23680-84-4
EINECS: 245-821-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR