2-Acetylphenylboronic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR10312-1G
OR10312-5G
OR10312-25G
APOSOR10312-1GEA
1410
CZK
APOSOR10312-1G
APOSOR10312-5G
APOSOR10312-25G
2-Acetylphenylboronic acid 95+%
2-Acetylphenylboronic acid
Vzorec:
C₈H₉BO₃ Storage Temperature: Freezer |
MDL Number:
MFCD01321263 CAS číslo: 308103-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...