(2-Benzothiazolylthio)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
[(1,3-Benzothiazol-2-yl)sulphanyl]acetic acid, 2-(Benzo[d]thiazol-2-ylthio)acetic acid
OR102929-1G
OR102929-5G
APOSOR102929-1GEA
622
CZK
APOSOR102929-1G
APOSOR102929-5G
(2-Benzothiazolylthio)acetic acid
(2-Benzothiazolylthio)acetic acid
Vzorec:
C₉H₇NO₂S₂ MW: 225,28 g/mol Bod tání: 159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00227391 CAS číslo: 6295-57-4 EINECS: 228-565-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...