Tisk…

5-Chlorobenzofuran-2-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chlorobenzo[b]furan-2-carboxylic acid

OR102841-25G OR102841-5G
APOSOR102841-25GEA 17000 CZK
APOSOR102841-25G APOSOR102841-5G
5-Chlorobenzofuran-2-carboxylic acid 97%
5-Chlorobenzofuran-2-carboxylic acid
Vzorec: C₉H₅ClO₃
MW: 196,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060512
CAS číslo: 10242-10-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR