5-Chloro-2,3,3-trimethyl-3H-indole
Dodavatel: Apollo Scientific
|
Varování
|
OR10207-25G
OR10207-100G
APOSOR10207-25GEA
4860
CZK
APOSOR10207-25G
APOSOR10207-100G
5-Chloro-2,3,3-trimethyl-3H-indole
5-Chloro-2,3,3-trimethyl-3H-indole
Vzorec:
C₁₁H₁₂ClN MW: 193.68 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00126966 CAS číslo: 25981-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...