Thiophene-2-ethylamine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene
OR10144-100G
OR10144-25G
APOSOR10144-100GEA
1540
CZK
APOSOR10144-100G
APOSOR10144-25G
Thiophene-2-ethylamine
Thiophene-2-ethylamine
Vzorec:
C₆H₉NS MW: 127,21 g/mol Bod varu: 201 °C Hustota: 1,087 g/cm³ (25 °C) Bod vzplanutí: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051495 CAS číslo: 30433-91-1 EINECS: 250-196-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...