Tisk…

2-(Chloromethyl)imidazo[1,2-a]pyridine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR1013-1G OR1013-5G
APOSOR1013-1GEA 6220 CZK
APOSOR1013-1G APOSOR1013-5G
2-(Chloromethyl)imidazo[1,2-a]pyridine hydrochloride
2-(Chloromethyl)imidazo[1,2-a]pyridine hydrochloride
Vzorec: C₈H₈Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD03094615
CAS číslo: 112230-20-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR