(Dimethylamino)acetaldehyde dimethylacetal 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,2-Dimethoxy-N,N-dimethylethanamine, N-(2,2-Dimethoxyethyl)dimethylamine
OR10113-5ML
APOSOR10113-5MLEA
622
CZK
APOSOR10113-5ML
(Dimethylamino)acetaldehyde dimethylacetal 98%
(Dimethylamino)acetaldehyde dimethylacetal
Vzorec:
C₆H₁₅NO₂ MW: 133,1888 g/mol Bod varu: 64…66 °C/12mmHg Hustota: 0,9 g/cm³ (25 °C) Bod vzplanutí: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010205 CAS číslo: 38711-20-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...