N,N-Diisopropylethylenediamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Aminoethyldiisopropylamine, N,N-Bis(isopropyl)ethane-1,2-diamine
OR10111-100G
OR10111-25G
OR10111-5G
APOSOR10111-100GEA
1740
CZK
APOSOR10111-100G
APOSOR10111-25G
APOSOR10111-5G
N,N-Diisopropylethylenediamine 98%
N,N-Diisopropylethylenediamine
Vzorec:
C₈H₂₀N₂ Bod varu: 170 °C Hustota: 0,83 g/cm³ (25 °C) Bod vzplanutí: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015015 CAS číslo: 121-05-1 EINECS: 204-447-4 UN: 2734 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...