Ethyl-7-bromoheptanoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-Bromoheptanoic Acid Ethyl Ester, Ethyl 7-bromoheptanoate
OR10083-100G
OR10083-25G
OR10083-5G
APOSOR10083-100GEA
6950
CZK
APOSOR10083-100G
APOSOR10083-25G
APOSOR10083-5G
Ethyl-7-bromoheptanoate
Ethyl-7-bromoheptanoate
Vzorec:
C₉H₁₇BrO₂ MW: 237.14 g/mol Bod varu: 112 °C/5mmHg Hustota: 1.217 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037103 CAS číslo: 29823-18-5 EINECS: 249-883-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...