N-Methyl-tert-butylamine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR10027-5G
OR10027-1G
OR10027-250MG
APOSOR10027-5GEA
4960
CZK
APOSOR10027-5G
APOSOR10027-1G
APOSOR10027-250MG
N-Methyl-tert-butylamine
N-Methyl-tert-butylamine
Vzorec:
C₅H₁₃N MW: 87,166 g/mol Bod varu: 67…69 °C Hustota: 0,727 g/cm³ (25 °C) Bod vzplanutí: –26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042853 CAS číslo: 14610-37-8 EINECS: 238-646-2 UN: 2733 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...