1-Phenylcyclopentanecarboxylic acid 99%
Dodavatel: Apollo Scientific
OR0993-100G
OR0993-25G
OR0993-5G
APOSOR0993-100GEA
900
CZK
APOSOR0993-100G
APOSOR0993-25G
APOSOR0993-5G
1-Phenylcyclopentanecarboxylic acid 99%
1-Phenylcyclopentanecarboxylic acid
Vzorec:
C₁₂H₁₄O₂ Bod tání: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001372 CAS číslo: 77-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...