Tisk…

1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride

OR0989-100G OR0989-25G OR0989-5G
APOSOR0989-100GEA 5960 CZK
APOSOR0989-100G APOSOR0989-25G APOSOR0989-5G
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride
Vzorec: C₈H₁₆ClN
MW: 198,13 g/mol
Bod tání: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00012842
CAS číslo: 26487-67-2
EINECS: 247-733-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR