Tisk…

1-Thiazol-2-ylpiperazine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(1-Piperazinyl)thiazole, 2-(Piperazin-1-yl)thiazole, 1-(2-Thiazolyl)piperazine

OR0960-1G OR0960-250MG OR0960-5G
APOSOR0960-1GEA 1160 CZK
APOSOR0960-1G APOSOR0960-250MG APOSOR0960-5G
1-Thiazol-2-ylpiperazine
1-Thiazol-2-ylpiperazine
Vzorec: C₇H₁₁N₃S
MW: 169,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00778419
CAS číslo: 42270-37-1
EINECS: 255-743-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR