Tisk…

3-Amino-5-nitrobenzoic acid 98%

Dodavatel: Apollo Scientific

OR0942-25G
APOSOR0942-25GEA 3940 CZK
APOSOR0942-25G
3-Amino-5-nitrobenzoic acid 98%
3-Amino-5-nitrobenzoic acid
Vzorec: C₇H₆N₂O₄
MW: 182,14 g/mol
Bod tání: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00017025
CAS číslo: 618-84-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR