4-Methoxy-3-nitroaniline
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR0934-25G
OR0934-1G
APOSOR0934-25GEA
1540
CZK
APOSOR0934-25G
APOSOR0934-1G
4-Methoxy-3-nitroaniline
4-Methoxy-3-nitroaniline
Vzorec:
C₇H₈N₂O₃ MW: 168.1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134138 CAS číslo: 577-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...