1H-Pyrazolo[3,4-b]pyridin-3-amine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1H-pyrazolo[3,4-b]pyridin-3-amine, 3-Amino-1H-pyrazolo[3,4-b]pyridine
OR0926-1G
OR0926-25G
OR0926-5G
APOSOR0926-1GEA
622
CZK
APOSOR0926-1G
APOSOR0926-25G
APOSOR0926-5G
1H-Pyrazolo[3,4-b]pyridin-3-amine
1H-Pyrazolo[3,4-b]pyridin-3-amine
Vzorec:
C₆H₆N₄ MW: 134,14 g/mol Bod tání: 187…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD04113611 CAS číslo: 6752-16-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...