Methyl-4-bromo-3-methylbenzoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-bromo-3-methylbenzoate
OR0924-5G
OR0924-25G
OR0924-1G
APOSOR0924-5GEA
705
CZK
APOSOR0924-5G
APOSOR0924-25G
APOSOR0924-1G
Methyl-4-bromo-3-methylbenzoate
Methyl-4-bromo-3-methylbenzoate
Vzorec:
C₉H₉BrO₂ MW: 229,07 g/mol Bod tání: 38…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673014 CAS číslo: 148547-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...