Diisopropyl azodicarboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Diisopropyl diazene-1,2-dicarboxylate
OR0923-100G
OR0923-25G
OR0923-500G
APOSOR0923-100GEA
992
CZK
APOSOR0923-100G
APOSOR0923-25G
APOSOR0923-500G
Diisopropyl azodicarboxylate 98%
Diisopropyl azodicarboxylate
Vzorec:
C₈H₁₄N₂O₄ Bod varu: 74…75 °C/0,3mmHg Bod tání: 44319 °C Hustota: 1,027 g/cm³ (25 °C) Bod vzplanutí: 106 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008875 CAS číslo: 2446-83-5 EINECS: 219-502-8 UN: 3082 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...