3-Methylpicolinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Methylpyridine-2-carboxylic acid
OR0922-1G
OR0922-5G
APOSOR0922-1GEA
622
CZK
APOSOR0922-1G
APOSOR0922-5G
3-Methylpicolinic acid
3-Methylpicolinic acid
Vzorec:
C₇H₇NO₂ Bod tání: 114…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191553 CAS číslo: 4021-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...