Tisk…

(4-(Bromomethyl)phenyl)acetic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Bromomethyl)phenylacetic acid

OR0901-25G OR0901-5G
APOSOR0901-25GEA 1740 CZK
APOSOR0901-25G APOSOR0901-5G
(4-(Bromomethyl)phenyl)acetic acid 97%
(4-(Bromomethyl)phenyl)acetic acid
Vzorec: (C₆H₄(CH₂Br))CH₂COOH
MW: 229,07 g/mol
Bod tání: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00010632
CAS číslo: 13737-36-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR