Tisk…

5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde

Dodavatel: Apollo Scientific

Synonyms: 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carbaldehyde, 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde, 5-(4-Hydroxypiperidino)thiophene-2-carboxaldehyde

OR0900-5G
APOSOR0900-5GEA 6520 CZK
APOSOR0900-5G
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde
Vzorec: C₁₀H₁₃NO₂S
MW: 211,28 g/mol
Bod tání: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD03425728
CAS číslo: 207290-72-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR