2-(1H-Imidazol-1-yl)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Imidazoleacetic acid, Imidazol-1-yl-acetic acid, 1-(Carboxymethyl)imidazole, 2-(1-Imidazolyl)acetic acid
OR0882-100G
OR0882-500G
APOSOR0882-100GEA
1620
CZK
APOSOR0882-100G
APOSOR0882-500G
2-(1H-Imidazol-1-yl)acetic acid
2-(1H-Imidazol-1-yl)acetic acid
Vzorec:
C₅H₆N₂O₂ MW: 126,1133 g/mol Bod tání: 255…257 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD01721800 CAS číslo: 22884-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...