4-Hydroxybenzylamine hydrobromide 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR0853-5G
OR0853-25G
APOSOR0853-5GEA
2440
CZK
APOSOR0853-5G
APOSOR0853-25G
4-Hydroxybenzylamine hydrobromide 97%
4-Hydroxybenzylamine hydrobromide
Vzorec:
C₇H₁₀BrNO Bod tání: 200…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258769 CAS číslo: 90430-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...