2,2'-Thiodiacetic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Thiodiglycolic acid, Thiodiacetic Acid
OR0788-100G
OR0788-1KG
OR0788-500G
APOSOR0788-100GEA
827
CZK
APOSOR0788-100G
APOSOR0788-1KG
APOSOR0788-500G
2,2'-Thiodiacetic acid 98%
2,2'-Thiodiacetic acid
Vzorec:
C₄H₆O₄S MW: 150.16 Bod varu: 168 Bod tání: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004359 CAS číslo: 123-93-3 EINECS: 204-663-9 UN: 3261 ADR: 8 II Merck Index: 12,09467 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...