2-Amino-4-chlorothiophenol 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-4-chlorobenzenethiol, 2-Amino-4-chlorophenyl mercaptan, 5-Chloro-2-mercaptoaniline
OR0785-25G
OR0785-5G
APOSOR0785-25GEA
1290
CZK
APOSOR0785-25G
APOSOR0785-5G
2-Amino-4-chlorothiophenol 97%
2-Amino-4-chlorothiophenol
Vzorec:
C₆H₆ClNS MW: 159,64 g/mol Bod tání: 56…61 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00792528 CAS číslo: 1004-00-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...