Bis(3-nitrophenyl)disulphide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,3'-Dinitrodiphenyl disulphide
OR0768-10G
OR0768-1G
OR0768-5G
APOSOR0768-10GEA
5590
CZK
APOSOR0768-10G
APOSOR0768-1G
APOSOR0768-5G
Bis(3-nitrophenyl)disulphide
Bis(3-nitrophenyl)disulphide
Vzorec:
C₁₂H₈N₂O₄S₂ MW: 308,34 g/mol Bod tání: 78…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007246 CAS číslo: 537-91-7 EINECS: 208-677-6 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...