Toluene-p-sulphonic anhydride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride
OR0751-100G
OR0751-25G
OR0751-500G
APOSOR0751-100GEA
1240
CZK
APOSOR0751-100G
APOSOR0751-25G
APOSOR0751-500G
Toluene-p-sulphonic anhydride 95%
Toluene-p-sulphonic anhydride
Vzorec:
C₁₄H₁₄O₅S₂ Bod tání: 121…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008548 CAS číslo: 4124-41-8 EINECS: 223-926-9 UN: 2585 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...