3-Nitro-4-(1-piperidinyl)benzaldehyde 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR0682-100MG
OR0682-10G
OR0682-1G
APOSOR0682-100MGEA
622
CZK
APOSOR0682-100MG
APOSOR0682-10G
APOSOR0682-1G
3-Nitro-4-(1-piperidinyl)benzaldehyde 95+%
3-Nitro-4-(1-piperidinyl)benzaldehyde
Vzorec:
C₁₂H₁₄N₂O₃ MW: 234,26 g/mol Bod tání: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00831780 CAS číslo: 39911-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...