Tisk…

6-Methylquinoxaline

Dodavatel: Apollo Scientific

OR0653-5G OR0653-100G OR0653-25G
APOSOR0653-5GEA 0 CZK
APOSOR0653-5G APOSOR0653-100G APOSOR0653-25G
6-Methylquinoxaline
6-Methylquinoxaline
Vzorec: C₉H₈N₂
MW: 144,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041001
CAS číslo: 6344-72-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR