1-Methyl-4-piperidone 98+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
N-Methyl-4-piperidone
OR0648-25G
OR0648-100G
OR0648-5G
APOSOR0648-25GEA
1780
CZK
APOSOR0648-25G
APOSOR0648-100G
APOSOR0648-5G
1-Methyl-4-piperidone 98+%
1-Methyl-4-piperidone
Vzorec:
C₆H₁₁NO MW: 113,16 g/mol Bod varu: 55…58 °C/10mmHg Hustota: 0,973 g/cm³ (25 °C) Bod vzplanutí: 58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006191 CAS číslo: 1445-73-4 EINECS: 215-895-5 UN: 2733 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...