Tisk…

3-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-propanol 95+%

Dodavatel: Apollo Scientific

Synonyms: Fmoc-β-Ala-ol, 3-(Fmoc-amino)-1-propanol

OR0487-100G OR0487-25G
APOSOR0487-100GEA 7910 CZK
APOSOR0487-100G APOSOR0487-25G
3-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-propanol 95+%
3-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-propanol
Vzorec: C₁₈H₁₉NO₃
MW: 297,35 g/mol
Bod tání: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00409484
CAS číslo: 157887-82-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR