Ethyl diethoxyacetate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Diethoxyacetic ethyl ester
OR0457-100G
OR0457-25G
OR0457-500G
APOSOR0457-100GEA
1120
CZK
APOSOR0457-100G
APOSOR0457-25G
APOSOR0457-500G
Ethyl diethoxyacetate 97%
Ethyl diethoxyacetate
Vzorec:
C₈H₁₆O₄ MW: 176.21 Bod varu: 198…200 °C Hustota: 0.985 g/cm³ (25 °C) Bod vzplanutí: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009231 CAS číslo: 6065-82-3 EINECS: 227-999-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...