2-Thiazolecarbonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1,3-Thiazole-2-carbonyl chloride, 1,3-Thiazole-2-carbonylchloride
OR0455-1G
OR0455-250MG
OR0455-500MG
APOSOR0455-1GEA
6720
CZK
APOSOR0455-1G
APOSOR0455-250MG
APOSOR0455-500MG
2-Thiazolecarbonyl chloride 97%
2-Thiazolecarbonyl chloride
Vzorec:
C₄H₂ClNOS MW: 147,582 g/mol Bod tání: 54…58 °C Storage Temperature: Freezer |
MDL Number:
MFCD06658970 CAS číslo: 30216-57-0 UN: 3261 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...