2-Ethoxyphenyl isocyanate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Ethoxy-2-isocyanatobenzene
OR0434-25G
OR0434-5G
APOSOR0434-25GEA
7280
CZK
APOSOR0434-25G
APOSOR0434-5G
2-Ethoxyphenyl isocyanate 97%
2-Ethoxyphenyl isocyanate
- May form small amount of sediment.
Vzorec:
C₉H₉NO₂ Bod varu: 119…121 °C/22mmHg Hustota: 1,105 g/cm³ (25 °C) Bod vzplanutí: 102 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002007 CAS číslo: 5395-71-1 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...