Tisk…

2,3-Dihydro-1-benzofuran-7-carbonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR0421-1G OR0421-250MG OR0421-500MG
APOSOR0421-1GEA 17860 CZK
APOSOR0421-1G APOSOR0421-250MG APOSOR0421-500MG
2,3-Dihydro-1-benzofuran-7-carbonyl chloride
2,3-Dihydro-1-benzofuran-7-carbonyl chloride
Vzorec: C₉H₇ClO₂
MW: 182,61 g/mol
Bod tání: 55…61 °C
Storage Temperature: Ambient
MDL Number: MFCD06658967
CAS číslo: 123266-63-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR