3,5-Dimethylphenyl isocyanate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,5-Xylyl isocyanate, 3,5-Dimethylphenylisocyanate
OR0415-25G
OR0415-5G
OR0415-100G
APOSOR0415-25GEA
2570
CZK
APOSOR0415-25G
APOSOR0415-5G
APOSOR0415-100G
3,5-Dimethylphenyl isocyanate
3,5-Dimethylphenyl isocyanate
- May form small amount of sediment.
Vzorec:
C₉H₉NO MW: 147,18 g/mol Bod varu: 205 °C Hustota: 1,045 g/cm³ (25 °C) Bod vzplanutí: 83 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013868 CAS číslo: 54132-75-1 EINECS: 258-987-0 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...