Tisk…

2,4-Dichloro-6-methylbenzenesulphonamide

Dodavatel: Apollo Scientific

OR0353-1G OR0353-5G OR0353-10G
APOSOR0353-1GEA 2980 CZK
APOSOR0353-1G APOSOR0353-5G APOSOR0353-10G
2,4-Dichloro-6-methylbenzenesulphonamide
2,4-Dichloro-6-methylbenzenesulphonamide
Vzorec: C₇H₇Cl₂NO₂S
Bod tání: 178…180 °C
Storage Temperature: Ambient
CAS číslo: 568577-80-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR