Tisk…

2-Chloro-4-(methylsulphonyl)benzoic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid

OR0264-100G OR0264-25G OR0264-5G
APOSOR0264-100GEA 953 CZK
APOSOR0264-100G APOSOR0264-25G APOSOR0264-5G
2-Chloro-4-(methylsulphonyl)benzoic acid 97%
2-Chloro-4-(methylsulphonyl)benzoic acid
Vzorec: C₈H₇ClO₄S
MW: 234,66 g/mol
Bod tání: 191…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00216496
CAS číslo: 53250-83-2
EINECS: 406-520-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR