Tisk…

4-Picolyl chloride hydrochloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride

OR0263-100G OR0263-25G OR0263-500G
APOSOR0263-100GEA 1660 CZK
APOSOR0263-100G APOSOR0263-25G APOSOR0263-500G
4-Picolyl chloride hydrochloride
4-Picolyl chloride hydrochloride
Vzorec: C₆H₇Cl₂N
MW: 164,03 g/mol
Bod tání: 166…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012826
CAS číslo: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR